Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585348 Supplier Invitrogen™ Supplier No. PA585348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (98%), Pig (100%), Chicken (100%), Sheep (100%), Rhesus Monkey (100%). Positive Control: MCF-7, MDA-MB-231, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF6
Applications Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ARF6
Gene Accession No. P62330, P62331, P62332
Gene Alias ADP ribosylation factor 6; ADP-ribosylation factor 6; AI788669; ARF6; AW496366; DKFZp564M0264
Gene Symbols ARF6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARF6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11845, 382, 79121
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.