Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF6 Rabbit anti-Human, Clone: JF101-2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267165
Description
ARF6 Monoclonal specifically detects ARF6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ARF6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| ADP-ribosylation factor 6, DKFZp564M0264 | |
| Rabbit | |
| Protein A purified | |
| Cell Cycle and Replication, Signal Transduction | |
| 382 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| JF101-2 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| ARF6 | |
| Synthetic peptide within Human ARF6 aa 133-165/ 175. (SwissProt: P62330 Human; SwissProt: P62331 Mouse; SwissProt: P62332 Rat) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction