Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF6 Rabbit anti-Human, Mouse, Rat, Clone: 9E7T9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | ARF6 |
---|---|
Clone | 9E7T9 |
Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
ARF6 Monoclonal antibody specifically detects ARF6 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ARF6 | |
Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ADP-ribosylation factor 6, DKFZp564M0264 | |
A synthetic peptide corresponding to a sequence within amino acids 76-175 of human ARF6 (P62330). HYYTGTQGLIFVVDCADRDRIDEARQELHRIINDREMRDAIILIFANKQDLPDAMKPHEIQEKLGLTRIRDRNWYVQPSCATSGDGLYEGLTWLTSNYKS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
9E7T9 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
382 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title