Learn More
Invitrogen™ ARF7 Monoclonal Antibody (2C8)

Description
Peptide Sequence: MGSLGSKNPQ TKQAQVLLLG LDSAGKSTLL YKLKLAKDIT TIPTIGFNVE MIELERNFSL TVWDVGGQEK MRTVWGCYCE NTDGLVYVVD STDKQRLEES QRQFEHILKN EHIKNVPVVL LANKQDMPGA LTAEDITRMF KVKKLCSDRN WYVQPCCALT GEGLAQGFRK LTGFVKSHMK SRGDTLAFFK QN.
Specifications
Specifications
| Antigen | ARF7 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2C8 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ARL14 |
| Gene Accession No. | Q8N4G2 |
| Gene Alias | 9130014L17Rik; ADP ribosylation factor like GTPase 14; ADP-ribosylation factor 7; ADP-ribosylation factor like GTPase 14; ADP-ribosylation factor-like 14; ADP-ribosylation factor-like protein 14; Arf7; Arl14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.