Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arfaptin 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983269
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3269 Supplier Bioss Supplier No. BS1442R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Putative target protein of ADP-ribosylation factor. Involved in membrane ruffling.

Specifications

Antigen Arfaptin 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARFIP2
Gene Accession No. P53365
Gene Alias 2310002N04Rik; ADP ribosylation factor interacting protein 2; ADP-ribosylation factor interacting protein 2; ADP-ribosylation factor interacting protein 2 (arfaptin 2); ADP-ribosylation factor-interacting protein 2; Arfaptin 2; Arfaptin-2; Arfip2; Partner of RAC1; partner of RAC1 (arfaptin 2); POR; POR1; Protein POR1
Gene Symbols ARFIP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Arfaptin 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23647
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.