Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARFGAP1 Antibody (1C4E2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261670
Description
ARFGAP1 Monoclonal specifically detects ARFGAP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
ARFGAP1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924 | |
Mouse | |
44.7 kDa | |
0.1 mL | |
55738 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Flow Cytometry, ELISA | |
1C4E2 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ARFGAP1 | |
Purified recombinant fragment of human ARFGAP1 (AA: 270-414) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction