Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARFGAP1 Antibody (OTI1F6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272013PE
Description
ARFGAP1 Monoclonal specifically detects ARFGAP1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
ARFGAP1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ARFGAP1 | |
Full length recombinant protein of human ARFGAP1 (NP_783202) produced in HEK293T cells. | |
0.1 mL | |
55738.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1F6 | |
Flow Cytometry | |
ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction