Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARFGAP1 Monoclonal Antibody (OTI2C5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502626S


Description
ARFGAP1 Monoclonal antibody specifically detects ARFGAP1 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
ARFGAP1 | |
Monoclonal | |
0.53 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P60981 | |
Mouse | |
Affinity chromatography | |
RUO | |
11034, 477147 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2C5 | |
Unconjugated | |
DSTN | |
ADP ribosylation factor GTPase activating protein 1; ADP-ribosylation factor 1 GTPase activating protein; ADP-ribosylation factor 1 GTPase-activating protein; ADP-ribosylation factor GTPase activating protein 1; ADP-ribosylation factor GTPase-activating protein 1; AI115377; ARF GAP 1; Arf GAP1; ARF1 GAP; ARF1-directed GTPase-activating protein; Arf1gap; ARFGAP1; EGK_02194; GAP protein; HRIHFB2281; RP11-261N11.1 | |
Full length human recombinant protein of human DSTN produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction