Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFGAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553466 Supplier Invitrogen™ Supplier No. PA553466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDNMSSAVPN HSPEKKDSDF FTEHTQPPAW DAPATEPSGT QQPAPSTESS GLAQPEHGPN TDLLGTSPKA SLELKSSIIG KKKPAAAKKG LGAKKGLGAQ KVSSQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

ARFGAP2 gene ontology annotations related to this gene include GTPase activator activity; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arfgap2
Gene Accession No. Q8N6H7
Gene Alias 2310032E02Rik; ADP ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase-activating protein 2; ARF GAP 2; ARFGAP2; GTPase-activating protein ZNF289; IRZ; NBLA10535; Zfp289; Zinc finger protein 289; zinc finger protein 289, ID1 regulated; Znf289
Gene Symbols Arfgap2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARFGAP2. Recombinant protein control fragment (Product #RP-91644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84364
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.