Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFGAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581884 Supplier Invitrogen™ Supplier No. PA581884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKQDILTIFK RLRSVPTNKV CFDCGAKNPS WASITYGVFL CIDCSGSHRS LGVHLSFIRS TELDSNWSWF QLRCMQVGGN ASASSFFHQH GCSTNDTNAK YNSRAAQLYR EKIKSLASQA TRKHGTDLWL DSCVVPPLSP PP Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.

The protein encoded by this gene is a GTPase-activating protein (GAP) that associates with the Golgi apparatus and regulates the early secretory pathway of proteins. The encoded protein promotes hydrolysis of ADP-ribosylation factor 1 (ARF1)-bound GTP, which is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is a prerequisite for the fusion of these vesicles with target compartments. The activity of this protein is sensitive to phospholipids. Multiple transcript variants encoding different isoforms have been found for this gene. This gene was originally known as ARFGAP1, but that is now the name of a related but different gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGAP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARFGAP3
Gene Accession No. Q9NP61
Gene Alias 0610009H19Rik; 1810004P07Rik; 1810035F16Rik; 9130416J18Rik; ADP ribosylation factor GTPase activating protein 3; ADP-ribosylation factor GTPase activating protein 1; ADP-ribosylation factor GTPase activating protein 3; ADP-ribosylation factor GTPase-activating protein 3; ARF GAP 3; ARF1GAP; ARFGAP1; ARFGAP3; arfgap3 {ECO:0000250; FLJ45618; HRIHFB2281; MGC39924; OTTHUMP00000028526; RP11-261N11.1; RP1-47A17.3; UniProtKB:Q9NP61}
Gene Symbols ARFGAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARFGAP3. Recombinant protein control fragment (Product #RP-91400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26286
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.