Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFGEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554790 Supplier Invitrogen™ Supplier No. PA554790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLYDGENHDC EEKPQDIVQN IVEEMVNIVV GDMGEGTTIN ASADGNIGTI EDGSDSENIQ ANGIPGTPIS VAYTPSLPDD RLSVSSNDTQ ESGNSSGPSP GAKF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGEF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARFGEF1
Gene Accession No. Q9Y6D6
Gene Alias ADP ribosylation factor guanine nucleotide exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited); ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited); Arfgef1; ARFGEP1; BIG1; Brefeldin A-inhibited GEP 1; brefeldin A-inhibited guanine nucleotide-exchange protein 1; D130059B05Rik; D730028O18Rik; DKFZP434L057; guanine nucleotide-exchange protein; hm:zeh1651; im:7138204; LOC553514 protein; LOW QUALITY PROTEIN: brefeldin A-inhibited guanine nucleotide-exchange protein 1; P200; p200 ARF guanine nucleotide exchange factor; p200 ARF-GEP1; RGD1560793; wu:fj64h08; zeh1651
Gene Symbols ARFGEF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARFGEF1. Recombinant protein control fragment (Product #RP-93734).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.