Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARFGEF2/BIG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1682 Supplier Bioss Supplier No. BS11804R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq, Jul 2008].

Specifications

Antigen ARFGEF2/BIG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARFGEF2
Gene Accession No. Q9Y6D5
Gene Alias ADP ribosylation factor guanine nucleotide exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited); AI463430; ARFGEF2; ARFGEP2; AW493672; BIG2; Brefeldin A-inhibited GEP 2; Brefeldin A-inhibited guanine nucleotide-exchange factor 2; Brefeldin A-inhibited guanine nucleotide-exchange protein 2; dJ1164I10.1; E230011G24Rik; PVNH2
Gene Symbols ARFGEF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARFGEF2/BIG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10564
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.