Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFGEF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555287 Supplier Invitrogen™ Supplier No. PA555287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPIQSKPQSP VIQAAAVSPK FVRLKHSQAQ SKPTTPEKTD LTNGEHARSD SGKVSTENGD APRERGSSLS GTDDGAQEVV KDILEDVVTS AIKAAEKHGL TEPERVLGEL ECQECAIPPG VDENSQTNGI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 74%.

ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGEF2
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARFGEF2
Gene Accession No. Q9Y6D5
Gene Alias ADP ribosylation factor guanine nucleotide exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited); AI463430; Arfgef2; ARFGEP2; AW493672; Big2; Brefeldin A-inhibited GEP 2; Brefeldin A-inhibited guanine nucleotide-exchange factor 2; brefeldin A-inhibited guanine nucleotide-exchange protein 2; dJ1164I10.1; E230011G24Rik; PVNH2
Gene Symbols ARFGEF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARFGEF2. Recombinant protein control fragment (Product #RP-89661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10564
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.