Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Arformoterol Tartrate
GREENER_CHOICE

Catalog No. 621910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-191-0 10 mg
62-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-191-0 Supplier Tocris Bioscience™ Supplier No. 6219/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Low molecular weight, organic compound that may help regulate biological process.

Arformoterol tartrate is a long-acting β2 adrenergic agonist (LABA). Bronchodilator.

Specifications

CAS 200815-49-2
Quantity 10 mg
Target Adrenergic beta-2
Molecular Formula C19H24N2O4.C4H6O6
Formula Weight 494.49g/mol
Product Type Long-acting Agonist (LABA)

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.