Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARFRP1 Antibody (SR1362), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322541100UL
Description
ARFRP1 Monoclonal antibody specifically detects ARFRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ARFRP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
SR1362 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
ADP-ribosylation factor related protein 1, ARF-related protein 1, ARL18, ARP1, ARPADP-ribosylation factor-related protein 1, helicase-like protein NHL, SCG10 like-protein | |
A synthesized peptide derived from human ARFRP1 (Uniprot #: Q13795) | |
100 μg | |
Signal Transduction | |
10139 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction