Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550606 Supplier Invitrogen™ Supplier No. PA550606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ARFRP1 protein.

The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFRP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ARFRP1
Gene Accession No. Q13795, Q63055, Q8BXL7
Gene Alias 1500006I01Rik; ADP ribosylation factor related protein 1; ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; AI480700; ARF-related protein 1; Arfrp1; ARL18; ARP; Arp1; helicase-like protein NHL; SCG10 like-protein
Gene Symbols ARFRP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human ADP-ribosylation factor related protein 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10139, 117051, 76688
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.