Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARFRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565630 Supplier Invitrogen™ Supplier No. PA565630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSKTRFNKNYK GMSLSKITTT VGLNIGTVDV GKARLMFWDL GGQEELQSLW DKYYAECHGV IYVIDSTDEE RLAESKQAFE KVVTSE Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFRP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARFRP1
Gene Accession No. Q13795
Gene Alias 1500006I01Rik; ADP ribosylation factor related protein 1; ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; AI480700; ARF-related protein 1; Arfrp1; ARL18; ARP; Arp1; helicase-like protein NHL; SCG10 like-protein
Gene Symbols ARFRP1
Host Species Rabbit
Immunogen Recombinant Human ARFRP1. Recombinant protein control fragment (Product #RP-106299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.