Learn More
Invitrogen™ ARFRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120267
Description
Positive test controls include: U-87MG, 293T, Mouse brain, Mouse heart, Rat lung. The target is usually found in the following locations: Golgi apparatus, trans-Golgi network. Immunogen sequence: MYTLLSGLYK YMFQKDEYCI LILGLDNAGK TTFLEQSKTR FNKNYKGMSL SKITTTVGLN IGTVDVGKAR LMFWDLGGQE ELQSLWDKYY AECHGVIYVI DSTDEERLAE SKQAFEKVVT SEALCGVPVL VLANKQDVET CLSIPDIKTA FSDCTSKIGR RDCLTQACSA LTGKGVREGI EWMVKCVVRN VHRPPRQRDI T.
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
ARFRP1 | |
Polyclonal | |
Unconjugated | |
ARFRP1 | |
1500006I01Rik; ADP ribosylation factor related protein 1; ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; AI480700; ARF-related protein 1; Arfrp1; ARL18; ARP; Arp1; helicase-like protein NHL; SCG10 like-protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
10139, 117051, 76688 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q13795, Q63055, Q8BXL7 | |
ARFRP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human ARFRP1 (NP_001254476.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.