Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARG1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5D6D12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ARG1 |
---|---|
Clone | 5D6D12 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6565
![]() ![]() |
Proteintech Group Inc
661291IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Arginase-1 (ARG1) catalyzes the hydrolysis of L-arginine into ornithine and urea, a critical step for the urea cycle. Defects in ARG1 are the cause of argininemia, an autosomal recessive disorder characterized by hyperammonemia. There are several isoforms of mammalian arginase. The type I isoform, is a cytosolic enzyme and expressed predomitly in the liver as a component of the urea cycle. Hepatocellular carcinoma usually shows higher protein expression of ARG1 than normal liver cells.Specifications
ARG1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARG1, Arginase 1, liver Arginase, Liver type arginase | |
Arg1 | |
IgG1 | |
Protein G | |
Antibody |
5D6D12 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P05089, P07824, Q61176, Q95JC8 | |
11846, 29221, 383, 397115 | |
ARG1 Fusion Protein Ag8810 | |
Primary | |
-20°C | |
ARG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title