Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARG1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ADG-1, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19575
Description
ARG1 Monoclonal antibody specifically detects ARG1 in Human, Mouse, Rat samples. It is validated for IP, WB-Ce, IHC-P.Specifications
| ARG1 | |
| Monoclonal | |
| Unconjugated | |
| ARG1 | |
| A synthetic peptide corresponding to human ARG1 | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| ADG-1 | |
| arginase, liver | |
| Rabbit | |
| 100 μg | |
| 383 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction