Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARG2 Antibody (OTI3G5), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270205AF488
Description
ARG2 Monoclonal specifically detects ARG2 in Human, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ARG2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARG2 | |
| Full length recombinant protein of human ARG2 (NP_001163) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3G5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| arginase 2, arginase, type II, arginase-2, mitochondrial, EC 3.5.3, EC 3.5.3.1, kidney arginase, Kidney-type arginase, L-arginine amidinohydrolase, L-arginine ureahydrolase, nonhepatic arginase, Non-hepatic arginase, Type II arginase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 384.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction