Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARG2 Antibody (OTI3G5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270205B
Description
ARG2 Monoclonal specifically detects ARG2 in Human, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ARG2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ARG2 | |
Full length recombinant protein of human ARG2 (NP_001163) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Canine | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3G5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
arginase 2, arginase, type II, arginase-2, mitochondrial, EC 3.5.3, EC 3.5.3.1, kidney arginase, Kidney-type arginase, L-arginine amidinohydrolase, L-arginine ureahydrolase, nonhepatic arginase, Non-hepatic arginase, Type II arginase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
384.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction