Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ [Arg8]-Vasopressin, MP Biomedicals™
Click to view available options
Quantity:
500 μg
Description
AVP is implicated in a variety of physiological processes.
- Endogenous antidiuretic hormone that regulates water balance
- Affects rat renal medullary Na+/K+ ATPase by arginine vasopressin mediation through the V2 receptor
- Reported to act as a neurotransmitter in the central nervous system
- Contraction of arterioles (vasopressor action)

Specifications
Specifications
Molecular Weight (g/mol) | 1084.239 g/mol |
Name | [Arg8]-Vasopressin |
Quantity | 500 μg |
Storage Requirements | Store at -20°C |
Regulatory Status | RUO |
Form | Lyophilized |
Purity or Quality Grade | 95+% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction