Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARHGEF10L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555310 Supplier Invitrogen™ Supplier No. PA555310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATVHPTICLG LQDGSILLYS SVDTGTQCLV SCRSPGLQPV LCLRHSPFHL LAGLQDGTLA AYPRTSGGVL WDLESPPVCL TVGPGPVRTL LSLEDAVWAS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

ARHGEF10L (Rho Guanine Nucleotide Exchange Factor 10 Like) is a Protein Coding gene. Among its related pathways areRET signalingandG protein signaling_RhoB regulation pathway. Gene Ontology (GO) annotations related to this gene includeRho guanyl nucleotide exchange factor activity. An important paralog of this gene isARHGEF10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF10L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF10L
Gene Accession No. Q9HCE6
Gene Alias argal; Arhgef10l; GrinchGEF; KIAA1626; Rho guanine nucleotide exchange factor (GEF) 10-like; Rho guanine nucleotide exchange factor 10 like; rho guanine nucleotide exchange factor 10-like protein
Gene Symbols ARHGEF10L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARGAL. Recombinant protein control fragment (Product #RP-94016).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55160
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.