Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Argatroban
GREENER_CHOICE

Catalog No. 163710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-371-0 10 mg
16-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-371-0 Supplier Tocris Bioscience™ Supplier No. 1637/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent thrombin inhibitor

Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.

This compound is a mixture of 2 diastereomers (approximate ratio 65/35).

Specifications

Chemical Name or Material Argatroban
CAS 74863-84-6
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C23H36N6O5S
Formula Weight 508.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.