Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARGBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557800 Supplier Invitrogen™ Supplier No. PA557800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRDSPRAISF KNGWQMARQN AEIWSSTEET VSPKIKSRSC DDLLNDDCDS FPDPKVKSES MGSLLCEEDS KESCPMAWGS PYVPEVRSNG RSR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 84%.

Arg and c-Abl represent the mammalian members of the Abelson family of non-receptor protein-tyrosine kinases. They interact with the Arg/Abl binding proteins via the SH3 domains present in the carboxy end of the latter group of proteins. ARGBP2 is the sorbin and SH3 domain containing 2 protein. It has three C-terminal SH3 domains and an N-terminal sorbin homology(SoHo) domain that interacts with lipid raft proteins. The subcellular localization of this protein in epithelial and cardiac muscle cells suggests that it functions as an adapter protein to assemble signaling complexes in stress fibers, and that it is a potential link between Abl family kinases and the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARGBP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SORBS2
Gene Accession No. O94875
Gene Alias 2010203O03Rik; 9430041O17Rik; A530071H08; Arg binding protein 2; Arg/Abl-interacting protein 2; Arg/Abl-interacting protein ArgBP2; arg-binding protein 2; ARGBP2; KIAA0777; leucine-rich repeat extensin-like protein 5; mKIAA0777; nArgBP2; Neural ArgBP2; PRO0618; Sorbin; sorbin and SH3 domain containing 2; sorbin and SH3 domain-containing protein 2; SORBS2
Gene Symbols SORBS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARGBP2. Recombinant protein control fragment (Product #RP-95659).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8470
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.