Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARGFX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560214 Supplier Invitrogen™ Supplier No. PA560214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVPALYSDAY DISQIIELYN LPDENEISSS SFHCLYQYLS LTKYQVGGQG SSLSIFAGPA VGLSPAQTWP NM Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 26%.

Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the ARGFX homeobox gene family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARGFX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARGFX
Gene Accession No. A6NJG6
Gene Alias ARGFX; Arginine-fifty homeobox
Gene Symbols ARGFX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARGFX. Recombinant protein control fragment (Product #RP-99960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 503582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.