Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody (658922), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB58681PECY7
Description
Arginase 1/ARG1/liver Arginase Monoclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Flow CytometrySpecifications
Arginase 1/ARG1/liver Arginase | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
658922 | |
Flow Cytometry | |
ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase | |
E. coli-derived recombinant human Arginase 1/ARG1, Met1-Lys322, Accession # P05089 | |
0.1 mL | |
Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
383 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction