Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132731APC
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Knockdown Validated.Specifications
Arginase 1/ARG1/liver Arginase | |
Polyclonal | |
Flow Cytometry, KnockDown Validated | |
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
Rabbit | |
Affinity Purified | |
Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell | |
383 | |
Store at 4°C in the dark. |
Flow Cytometry, KnockDown | |
APC | |
PBS with 0.05% Sodium Azide | |
ARG1 | |
Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction