Learn More
Arginase 1/ARG1/liver Arginase Antibody - BSA Free, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 9 publications
Supplier:  Novus Biologicals NBP132731
Catalog No. NBP132731
Only null left
Add to Cart
Description
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications
Specifications
Arginase 1/ARG1/liver Arginase
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Rabbit
35 kDa
0.1 mg
Cellular Markers, Chromatin Research, Lipid and Metabolism
383
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
1.0 mg/ml
Western Blot 1:1000-1:50000, Simple Western, Flow Cytometry 5 - 10 ug/ml, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, Knockdown Validated
P05089
ARG1
Arginase 1/ARG1/liver Arginase Antibody made from full length human Arginase 1 Recombinant protein.
Affinity Purified
RUO
Primary
Human, Mouse, Rat
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only