Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: Novus Biologicals NB1005974025UG
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.Specifications
Arginase 1/ARG1/liver Arginase | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
Goat | |
37 kDa | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
0.5 mg/mL | |
Western Blot 0.05 - 0.1 μg/mL, Simple Western 5 μg/mL, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA 1:128000 | |
P07824 | |
ARG1 | |
Peptide with sequence C-NHKPETDYLKPPK corresponding to C-Terminus according to NP_058830.2. | |
Affinity Purified | |
RUO | |
383 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction