Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Chicken anti-Human, Mouse, Rat, Guinea Pig, Polyclonal, Novus Biologicals™

Chicken Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Arginase 1/ARG1/liver Arginase |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:500 |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Arginase 1/ARG1/liver Arginase Polyclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Arginase 1/ARG1/liver Arginase | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Chicken | |
Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. | |
383 | |
Primary | |
Store at -20C. |
Western Blot 1:1000, Immunohistochemistry 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Guinea Pig | |
ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase | |
Full-length recombinant rat arginase I. (UniProt: P07824) | |
IgY purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title