Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arginase 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531088 Supplier Invitrogen™ Supplier No. PA531088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3. Predicted reactivity: Mouse (90%), Rat (87%), Dog (83%), Rabbit (88%), Chicken (81%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Arginase catalyzes the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exists (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type II isoform encoded by this gene, is located in the mitochondria and expressed in extra-hepatic tissues, especially kidney. The physiologic role of this isoform is poorly understood; it is thought to play a role in nitric oxide and polyamine metabolism. Transcript variants of the type II gene resulting from the use of alternative polyadenylation sites have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arginase 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene ARG2
Gene Accession No. O08691, O08701, P78540
Gene Alias AII; AII type II arginase; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; nonhepatic arginase; non-hepatic arginase; type II arginase
Gene Symbols ARG2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARG2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11847, 29215, 384
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.