Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginine, USP, 98.5-101.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Arginine, USP is a semiessential amino acid that has an important part in wound healing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Chloride | 0.05% |
Ignition Residue | 0.3% |
Loss on Drying | 0.5% |
Packaging | Amber Glass Bottle |
Specific Rotation | +26.3° to +27.7° |
Sulfate | 0.03% |
Quantity | 100 g |
Percent Purity | 98.5 to 101.5% |
Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction