Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Argininosuccinate Lyase Antibody, DyLight 680 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299243FR
Description
Argininosuccinate Lyase Polyclonal antibody specifically detects Argininosuccinate Lyase in Human samples. It is validated for Western Blot, ELISASpecifications
Argininosuccinate Lyase | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
DyLight 680 | |
argininosuccinase, argininosuccinate lyase, Arginosuccinase, ASAL, EC 4.3.2.1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Argininosuccinate Lyase. | |
0.1 mL | |
metabolism | |
435 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction