Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Argininosuccinate Lyase Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299442PE
Description
Argininosuccinate Lyase Polyclonal antibody specifically detects Argininosuccinate Lyase in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Argininosuccinate Lyase | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
argininosuccinase, argininosuccinate lyase, Arginosuccinase, ASAL, EC 4.3.2.1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Argininosuccinate Lyase | |
0.1 mL | |
metabolism | |
435 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction