Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginyl tRNA synthetase Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297635UV
Description
Arginyl tRNA synthetase Polyclonal specifically detects Arginyl tRNA synthetase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Arginyl tRNA synthetase | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
Produced in rabbits immunized with E. coli-derived Human Arginyl tRNA synthetase fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
5917 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction