Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arginyl tRNA synthetase Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297635PE
Description
Arginyl tRNA synthetase Polyclonal antibody specifically detects Arginyl tRNA synthetase in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Arginyl tRNA synthetase | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
Produced in rabbits immunized with E. coli-derived Human Arginyl tRNA synthetase fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
5917 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction