Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARGLU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566041 Supplier Invitrogen™ Supplier No. PA566041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSRSTNTAVSR RERDRERASS PPDRIDIFGR TVSKRSSLDE KQKR Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

ARGLU1 belongs to the UPF0430 family. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARGLU1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARGLU1
Gene Accession No. Q9NWB6
Gene Alias 9430010O03Rik; arginine and glutamate rich 1; arginine and glutamate-rich protein 1; arginine and glutamate-rich protein 1-A; LOW QUALITY PROTEIN: arginine and glutamate-rich protein 1; ARGLU1; C130008N12; LOW QUALITY PROTEIN: arginine and glutamate-rich protein 1; RGD1310061; Unknown (protein for MGC:134421)
Gene Symbols ARGLU1
Host Species Rabbit
Immunogen Recombinant Human ARGLU1. Recombinant protein control fragment (Product #RP-105053).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.