Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550654 Supplier Invitrogen™ Supplier No. PA550654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total AGO1 protein.

AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. AGO1 binds to miRNAs or siRNAs and represses the translation of mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ago1
Gene Accession No. Q8CJG1, Q9UL18
Gene Alias AGO1; argonaute 1; argonaute 1, RISC catalytic component; argonaute RISC catalytic component 1; argonaute RISC catalytic subunit 1; argonaute1; EIF2C; eIF2C 1; eIF-2C 1; EIF2C1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; mAgo1; Piwi/Argonaute family protein meIF2C1; Protein argonaute-1; putative RNA-binding protein Q99; Q99
Gene Symbols Ago1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human argonaute RISC catalytic component 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236511, 26523
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.