Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Argonaute 3 Antibody (PCRP-AGO3-1C5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314202APC
This item is not returnable.
View return policy
Description
Argonaute 3 Monoclonal antibody specifically detects Argonaute 3 in Human samples. It is validated for Flow CytometrySpecifications
Argonaute 3 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
192669 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry | |
PCRP-AGO3-1C5 | |
Flow Cytometry | |
AGO3argonaute 3, argonaute3, eIF2C 3, eIF-2C 3, Eukaryotic translation initiation factor 2C 3, eukaryotic translation initiation factor 2C, 3, FLJ12765, hAgo3, MGC86946, protein argonaute-3 | |
Recombinant full-length human Argonaute 3 protein (Uniprot: Q9H9G7) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction