Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Argonaute 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567021 Supplier Invitrogen™ Supplier No. PA567021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQWVSVVSLQL LLEALAGHLN EVPDDSVQAL DVITRH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 57%.

This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Argonaute 4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGO4
Gene Accession No. Q9HCK5
Gene Alias 5730550L01Rik; AGO4; AI481660; argonaute 4; argonaute 4, RISC catalytic component; argonaute RISC catalytic component 4; argonaute RISC catalytic subunit 4; argonaute RISC component 4; argonaute4; eIF2C 4; eIF-2C 4; Eif2c4; Eukaryotic translation initiation factor 2C 4; eukaryotic translation initiation factor 2C, 4; hAgo4; KIAA1567; LOW QUALITY PROTEIN: protein argonaute-4; mAgo4; Piwi/Argonaute family protein meIF2C4; Protein argonaute-4
Gene Symbols AGO4
Host Species Rabbit
Immunogen Recombinant Human Argonaute 4. Recombinant protein control fragment (Product #RP-105460).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.