Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ Essentials BioSafe Serological Pipettes
Click to view available options
Volume (Metric):
5 mL
10 mL
25 mL
Graduations:
0.1 mL
0.2 mL
Description
- Eliminate cross-contamination - unique barrier filter prevents over-aspiration
- Polystyrene pipettes are sterile, individually wrapped, and disposable
- Ideal for use with potentially hazardous or dangerous biological fluids
- Use in place of filtered pipette tips
Specifications
Specifications
Accuracy | ±0.1 mL |
Aspirator Plug | Yes |
Autoclavable | Not Autoclavable |
Calibrated | Yes |
Calibration | To Deliver |
Material | Polystyrene |
Sterility | Sterile |
For Use With (Application) | Biological fluids |
Graduated | Yes |
Graduations | 0.1 mL |
Show More |
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction