Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials CELLine Bioreactor Flasks

Catalog No. p-3144285
Change view
Click to view available options
Capacity (Metric):
350 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-391-207 1000 mL
03-391-208 350 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-391-207 Supplier Cole-Parmer™ Essentials Supplier No. 2440653
Only null left
Add to Cart
Add to Cart

Ideal for monoclonal antibodies and expressed recombinant cellular protein production

  • Constructed with a polystyrene housing and molded silicone gas transfer bottom
  • Features a 10,000 MWCO regenerated cellulose semi-permeable membrane and 0.2μm bottom vent membrane barrier
  • Gamma irradiation sterilized and pressure tested

These bioreactor flasks are viable in vitro alternatives to using mouse ascites, standard cell culture, or tissue culture products. Bioreactors utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cells and cellular-secreted products with a MWCO over 10kD are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange. Separate ports give access to either media or cell cultivation chamber allowing easy change of expended medium without interference of cell growth, function, or gas exchange.

Specifications

Capacity (English) 33.8 oz.
Description 1 x 109 Cells/Cell Compartment
Capacity (Metric) 1000 mL
Material Polystyrene
For Use With (Application) For use with CHO, BHK, HEP2 or any other adherent cell line, for monoclonal Antibody production up to 5mg/mL
Product Type Bioreactor Flask
Shape Rectangular
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Mouth Style Threaded
Sterility Sterile
Bottom Shape Rectangular
Surface Treatment Non-treated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.