Learn More
Cole-Parmer™ Essentials CELLine Bioreactor Flasks
Description
- Constructed with a polystyrene housing and molded silicone gas transfer bottom
- Features a 10,000 MWCO regenerated cellulose semi-permeable membrane and 0.2μm bottom vent membrane barrier
- Gamma irradiation sterilized and pressure tested
These bioreactor flasks are viable in vitro alternatives to using mouse ascites, standard cell culture, or tissue culture products. Bioreactors utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cells and cellular-secreted products with a MWCO over 10kD are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange. Separate ports give access to either media or cell cultivation chamber allowing easy change of expended medium without interference of cell growth, function, or gas exchange.
Specifications
Specifications
| Capacity (English) | 33.8 oz. |
| Description | 1 x 109 Cells/Cell Compartment |
| Capacity (Metric) | 1000 mL |
| Material | Polystyrene |
| For Use With (Application) | For use with CHO, BHK, HEP2 or any other adherent cell line, for monoclonal Antibody production up to 5mg/mL |
| Product Type | Bioreactor Flask |
| Shape | Rectangular |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Graduated | Yes |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.