Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215449
Description
ARH Polyclonal specifically detects ARH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARH | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705 | |
Rabbit | |
34 kDa | |
0.1 mL | |
Lipid and Metabolism | |
26119 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q5SW96 | |
LDLRAP1 | |
Recombinant protein encompassing a sequence within the center region of human ARH. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Rat (83%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction