Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARH Antibody (OTI7E11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271790PE
Description
ARH Monoclonal specifically detects ARH in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
ARH | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
LDLRAP1 | |
Full length recombinant protein of human ARH (NP_056442) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI7E11 | |
Flow Cytometry | |
ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
26119.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction