Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDLRAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109904 Supplier Invitrogen™ Supplier No. PA5109904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQPKGEELSA AAIKRIVATA KASGKKLQKV TLKVSPRGII LTDNLTNQLI ENVSIYRISY CTADKMHDKV FAYIAQSQHN QSLECHAFLC TKRKMAQAVT LTVAQAFKVA FEFWQVSKEE KEKRDKASQE GGDVLGARQD CTPSLKSLVA TGNLLDLEET AKAPLSTVSA NTTNMDEVPR PQALSGSSVV WELDDGLDEA FSRLAQSRTN PQVLDTGLTA QDMHYAQCLS PVDWDKPDSS GTEQDDLFSF.

The protein encoded by this gene is a cytosolic protein which contains a phosphotyrosine binding (PTD) domain. The PTD domain has been found to interact with the cytoplasmic tail of the LDL receptor. Mutations in this gene lead to LDL receptor malfunction and cause the disorder autosomal recessive hypercholesterolaemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLRAP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LDLRAP1
Gene Accession No. D3ZAR1, Q5SW96, Q8C142
Gene Alias AA691260; Arh; Arh1; ARH2; Autosomal recessive hypercholesterolemia protein; autosomal recessive hypercholesterolemia protein homolog; DKFZp586D0624; FHCB1; FHCB2; LDL receptor adaptor protein; LDLRAP1; Low density lipoprotein receptor adapter protein 1; low density lipoprotein receptor adaptor protein 1; MGC34705; RGD1563417; RP11-70P17.2
Gene Symbols LDLRAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 59-308 of human LDLRAP1 (NP_056442.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100017, 26119, 500564
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.