Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARHE Monoclonal Antibody (5C7E8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531896
Description
MA5-31896 has been tested in indirect ELISA.
Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras, Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex; GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins; and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form.
Specifications
ARHE | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P61587 | |
RND3 | |
Purified recombinant fragment of human RND3 (AA: 104-241) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
5C7E8 | |
Unconjugated | |
RND3 | |
2610017M01Rik; AI661404; Arhe; memB; MemB protein; Protein MemB; ras homolog gene family, member E; Rho family GTPase 3; Rho related protein Rnd3/Rho8; rho8; RHOE; rho-related GTP-binding protein Rho8; Rho-related GTP-binding protein RhoE; Rnd3; small GTP binding protein Rho8 | |
Mouse | |
Protein G | |
RUO | |
390 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction