Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581251 Supplier Thermo Scientific Supplier No. PA581251
Add to Cart
Add to Cart

ARHE Polyclonal Antibody

ARHE Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHE
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RND3
Gene Accession No. P61587
Gene Alias 2610017M01Rik; AI661404; Arhe; memB; MemB protein; Protein MemB; ras homolog gene family, member E; Rho family GTPase 3; Rho related protein Rnd3/Rho8; rho8; RHOE; rho-related GTP-binding protein Rho8; Rho-related GTP-binding protein RhoE; Rnd3; small GTP binding protein Rho8
Gene Symbols RND3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human RND3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.